Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H3525

Sigma-Aldrich

6α-Hydroxytestosterone

Synonym(s):

4-Androstene-6α,17β-diol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic (organic)

form

solid

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

transformation: suitable

shipped in

ambient

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](O)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@@H](O)CC[C@@]24[H]

InChI

1S/C19H28O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,16-17,21-22H,3-8,10H2,1-2H3/t12-,13-,14-,16-,17-,18+,19-/m0/s1

InChI key

XSEGWEUVSZRCBC-QXROXWLYSA-N

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Liu et al.
Biological & pharmaceutical bulletin, 27(10), 1555-1560 (2004-10-07)
The intestinal bacterial metabolites of ginsenosides are responsible for the main pharmacological activities of ginseng. The purpose of this study was to find whether these metabolites influence hepatic metabolic enzymes and to predict the potential for ginseng-prescription drug interactions. Utilizing
Annalise Di Marco et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(3), 349-358 (2004-12-21)
A rapid and sensitive radiometric assay for assessing the potential of drugs to inhibit cytochrome P450 (P450) 3A4/5 in human liver microsomes is described. In contrast to the conventional testosterone 6beta-hydroxylation assay, the new method does not require high-performance liquid
Thomas M Polasek et al.
European journal of clinical pharmacology, 62(3), 203-208 (2006-01-18)
Macrolide antibiotics are mechanism-based inactivators of CYP3A enzymes that exhibit varying degrees of inhibitory potency. Our aim was to predict quantitatively the drug-drug interaction (DDI) potential of five macrolides from in vitro studies using testosterone as the CYP3A substrate, and
Dan A Rock et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2410-2413 (2008-09-04)
Cytochrome P450 (P450) reaction phenotyping is a key process toward accurately determining the contribution of different P450s to the metabolism of new chemical entities. The significance of P450s to drug disposition has led to the identification of selective chemical and
Tao Liu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(2), 254-259 (2012-07-25)
To study the influences of Shengbanfang on CYP3A1 activities of SD rat and provide suggestions for drug combinations. 25 male SD rats were devided into 5 groups randomly,and treated with saline( NS, ig, 10 mg/(kg/d) ,qd,14 d), dexamethasone (DEX, ig

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service