Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H3003

Sigma-Aldrich

16α-Hydroxyandrosterone

powder

Synonym(s):

3α,16α-Dihydroxy-5α-androstan-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O3
CAS Number:
Molecular Weight:
306.44
UNSPSC Code:
12352200

form

powder

color

white

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service