G8005
sn-Glycero-3-phosphocholine 1:1 cadmium chloride adduct
~98%, crystalline
Synonym(s):
L-α-Glycerophosphorylcholine 1:1 cadmium chloride adduct from egg yolk
About This Item
Recommended Products
assay
~98%
form
crystalline
storage temp.
−20°C
SMILES string
Cl[Cd]Cl.C[N+](C)(C)CCOP([O-])(=O)OC[C@H](O)CO
InChI
1S/C8H20NO6P.Cd.2ClH/c1-9(2,3)4-5-14-16(12,13)15-7-8(11)6-10;;;/h8,10-11H,4-7H2,1-3H3;;2*1H/q;+2;;/p-2/t8-;;;/m1.../s1
InChI key
RCOWLNSSDOITMP-VFIYQRISSA-L
Looking for similar products? Visit Product Comparison Guide
Reconstitution
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service