G5394
(R)-(+)-Glycidol
Synonym(s):
(R)-(+)-Oxirane-2-methanol
About This Item
Recommended Products
refractive index
n20/D 1.43 (lit.)
bp
56-57 °C/11 mmHg (lit.)
density
1.116 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
OC[C@@H]1CO1
InChI
1S/C3H6O2/c4-1-3-2-5-3/h3-4H,1-2H2/t3-/m1/s1
InChI key
CTKINSOISVBQLD-GSVOUGTGSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Self-react. C - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
5.2 - Organic peroxides and self-reacting hazardous materials
wgk_germany
WGK 3
flash_point_f
161.6 °F
flash_point_c
72 °C
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
suitable for GCsuitable for GCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service