Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G1134

Sigma-Aldrich

Glyoxylic acid solution

50 % (w/w) in H2O

Synonym(s):

Formylformic acid, Oxoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCOOH
CAS Number:
Molecular Weight:
74.04
Beilstein/REAXYS Number:
741891
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

50 % (w/w) in H2O

storage temp.

2-8°C

SMILES string

OC(=O)C=O

InChI

1S/C2H2O3/c3-1-2(4)5/h1H,(H,4,5)

InChI key

HHLFWLYXYJOTON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Couvidat et al.
Environmental science & technology, 47(2), 914-922 (2012-11-30)
A molecular surrogate representation of secondary organic aerosol (SOA) formation is used to investigate the effect of aqueous-phase (in clouds and particles) chemical processing and wet deposition on SOA atmospheric concentrations. To that end, the hydrophilic/hydrophobic organic (H(2)O) model was
Joseph P Dexter et al.
The Journal of biological chemistry, 288(8), 5770-5778 (2012-11-30)
An important goal of systems biology is to develop quantitative models that explain how specific molecular features give rise to systems-level properties. Metabolic and regulatory pathways that contain multifunctional proteins are especially interesting to study from this perspective because they
Jade C S Chung et al.
Open biology, 3(1), 120131-120131 (2013-02-01)
Pseudomonas aeruginosa is an opportunistic human pathogen and a common cause of chronic infections in individuals with cystic fibrosis (CF). Oxygen limitation was recently reported to regulate the expression of a major virulence determinant in P. aeruginosa, the type III
Kathrin Schneider et al.
Journal of bacteriology, 194(12), 3144-3155 (2012-04-12)
Oxalate catabolism is conducted by phylogenetically diverse organisms, including Methylobacterium extorquens AM1. Here, we investigate the central metabolism of this alphaproteobacterium during growth on oxalate by using proteomics, mutant characterization, and (13)C-labeling experiments. Our results confirm that energy conservation proceeds
Kelly C Fahnoe et al.
PloS one, 7(12), e51732-e51732 (2012-12-15)
Antibacterial compounds that affect bacterial viability have traditionally been identified, confirmed, and characterized in standard laboratory media. The historical success of identifying new antibiotics via this route has justifiably established a traditional means of screening for new antimicrobials. The emergence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service