Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0394

Sigma-Aldrich

Growth Hormone Releasing Factor Bovine Fragment 1-29 amide

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C150H247N45O42S
CAS Number:
Molecular Weight:
3384.91
MDL number:
UNSPSC Code:
12352200

assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)Cc1ccc(O)cc1)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

Gene Information

Amino Acid Sequence

Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Asn-Arg-NH2

Biochem/physiol Actions

Amidated GRF fragment that is equipotent to GRF in releasing somatotropin from anterior pituitary.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Lefrançois et al.
Neuroendocrinology, 59(4), 363-370 (1994-04-01)
Previous structure-activity studies on growth-hormone-releasing factor (GRF) have mainly been carried out in pituitary cell culture assays. In such systems, the molecular features necessary to increase GRF receptor affinity cannot be fully distinguished from those that improve proteolytic resistance. To
V A Lance et al.
Biochemical and biophysical research communications, 119(1), 265-272 (1984-02-29)
Human pancreatic growth hormone releasing factor (1-29)-amide [hpGRF (1-29)-NH2] and the following analogs: [D-Tyr-1]-hpGRF(1-29)-NH2, [D-Ala-2]-hpGRF(1-29)-NH2, [D-Asp-3]-hpGRF(1-29)-NH2, and [N-Ac-Tyr-1]-hpGRF (1-29)-NH2 were synthesized using solid phase methodology and tested for their ability to stimulate growth hormone (GH) secretion in the rat and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service