All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H6F6N2
CAS Number:
Molecular Weight:
280.17
MDL number:
UNSPSC Code:
12352200
assay
≥95% (NMR)
form
solid
SMILES string
FC(F)(F)C1(C2CCC(C2)C1(C#N)C#N)C(F)(F)F
Biochem/physiol Actions
A potent non-competitive GABA antagonist in mammals, molluscs and insects; convulsant.
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 2 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service