F5786
Formamide
BioXtra, ≥99% (GC)
Synonym(s):
Amide C1, Formic amide
About This Item
Recommended Products
vapor density
1.55 (vs air)
vapor pressure
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
product line
BioXtra
assay
≥99% (GC)
autoignition temp.
932 °F
impurities
<0.0005% Phosphorus (P)
ign. residue
<0.1%
refractive index
n20/D 1.447 (lit.)
bp
210 °C (lit.)
mp
2-3 °C (lit.)
density
1.134 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%
cation traces
Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%
SMILES string
NC=O
InChI
1S/CH3NO/c2-1-3/h1H,(H2,2,3)
InChI key
ZHNUHDYFZUAESO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
related product
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Repr. 1B - STOT RE 2 Oral
target_organs
Blood
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 1
flash_point_f
305.6 °F
flash_point_c
152 °C
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service