F2768
Fast Red TR Salt hemi(zinc chloride) salt
practical grade
Synonym(s):
4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11
About This Item
Recommended Products
grade
practical grade
composition
Dye content, ~15%
storage temp.
−20°C
SMILES string
[Cl-].[Cl-].Cl[Zn]Cl.Cc1cc(Cl)ccc1[N+]#N.Cc2cc(Cl)ccc2[N+]#N
InChI
1S/2C7H6ClN2.4ClH.Zn/c2*1-5-4-6(8)2-3-7(5)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4
InChI key
RIIVFDKBXXMYRL-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service