Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F2130

Sigma-Aldrich

Iron(II) chloride tetrahydrate

≥99%

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
EC Number:
MDL number:
UNSPSC Code:
12352302

vapor pressure

10 mmHg ( 693 °C)

assay

≥99%

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service