Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F2004

Sigma-Aldrich

Ammonium formate

Synonym(s):

Formic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
Beilstein/REAXYS Number:
3625095
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

119-121 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne M Filppula et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(7), 1202-1209 (2014-04-10)
Previous studies have shown that several protein kinase inhibitors are time-dependent inhibitors of cytochrome P450 (CYP) 3A. We screened 14 kinase inhibitors for time-dependent inhibition of CYP2C8 and CYP3A. Amodiaquine N-deethylation and midazolam 1'-hydroxylation were used as marker reactions for
Therese Fagerqvist et al.
Journal of neurochemistry, 126(1), 131-144 (2013-02-01)
Inclusions of intraneuronal alpha-synuclein (α-synuclein) can be detected in brains of patients with Parkinson's disease and dementia with Lewy bodies. The aggregation of α-synuclein is a central feature of the disease pathogenesis. Among the different α-synuclein species, large oligomers/protofibrils have
Jo Philips et al.
Journal of contaminant hydrology, 147, 25-33 (2013-03-19)
Acidification due to microbial dechlorination of trichloroethene (TCE) can limit the bio-enhanced dissolution of TCE dense non-aqueous phase liquid (DNAPL). This study related the dissolution enhancement of a TCE DNAPL to the pH buffer capacity of the medium and the
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.
Todd A Lydic et al.
Methods (San Diego, Calif.), 87, 83-95 (2015-04-25)
There is an increasing recognition of the role that cancer cell derived exosomes play in intercellular signaling upon fusion or uptake with a target cell, including immune system evasion, tumor growth and metastasis. To date, however, although exosomal membrane and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service