About This Item
vapor density
9.8 (vs air)
assay
~99%
form
waxy solid
color
white
bp
220 °C/30 mmHg (lit.)
mp
35-37 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCCCCCC
InChI
1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3
InChI key
CBFCDTFDPHXCNY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service