Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9129

Sigma-Aldrich

Ethylene glycol

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein/REAXYS Number:
505945
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

autoignition temp.

752 °F

expl. lim.

15.3 %

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

target_organs

Kidney

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

239.0 °F - open cup

flash_point_c

115 °C - open cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service