E4507
Ethyl 3-ethoxyacrylate
≥97%
Synonym(s):
Ethyl 3-ethoxy-2-propenoate
About This Item
assay
≥97%
refractive index
n20/D 1.446 (lit.)
bp
195-196 °C (lit.)
density
0.998 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCO\C=C\C(=O)OCC
InChI
1S/C7H12O3/c1-3-9-6-5-7(8)10-4-2/h5-6H,3-4H2,1-2H3/b6-5+
InChI key
ITQFPVUDTFABDH-AATRIKPKSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
168.8 °F - closed cup
flash_point_c
76 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service