Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E3882

Sigma-Aldrich

[D-Ala2, D-Met5]-Enkephalinamide acetate salt

≥97% (HPLC)

Synonym(s):

Tyr-D-Ala-Gly-Phe-D-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38N6O6S
CAS Number:
Molecular Weight:
586.70
MDL number:
UNSPSC Code:
12352200

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)=O.CSCC[C@@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)[C@@H](C)NC(=O)[C@@H](N)Cc2ccc(O)cc2)C(N)=O

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced analgesic activity of D-Ala2 enkephalinamides following D-isomer substitutions at position five.
J D Belluzzi et al.
Life sciences, 23(2), 99-104 (1978-07-10)
Peptides.
Ling. N., et al. et al.
Proceedings of the Fifth American Peptide Symposium, 96-96 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service