Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E0254

Sigma-Aldrich

Ethylbenzene-d10

Synonym(s):

Decadeuteroethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD2CD3
CAS Number:
Molecular Weight:
116.23
Beilstein/REAXYS Number:
1960387
EC Number:
MDL number:
UNSPSC Code:
12352200

refractive index

n20/D 1.492 (lit.)

bp

134.6 °C (lit.)

mp

-95 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3/i1D3,2D2,3D,4D,5D,6D,7D

InChI key

YNQLUTRBYVCPMQ-CFTAVCBPSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

target_organs

hearing organs

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

59.0 °F - closed cup

flash_point_c

15.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service