Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9412

Sigma-Aldrich

(5S,6S)-Dihydroxy-(7E,9E,11Z,14Z)-eicosatetraenoic acid

≥90%, ~100 μg/mL in ethanol

Synonym(s):

(5S,6S)-DiHETE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥90%

form

liquid

concentration

~100 μg/mL in ethanol

functional group

carboxylic acid

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C/C=C\C=C\C=C\[C@H](O)[C@@H](O)CCCC(O)=O

InChI

1S/C20H32O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18(21)19(22)16-14-17-20(23)24/h6-7,9-13,15,18-19,21-22H,2-5,8,14,16-17H2,1H3,(H,23,24)/b7-6-,10-9-,12-11+,15-13+/t18-,19-/m0/s1

InChI key

UVZBUUTTYHTDRR-WAQVJNLQSA-N

Packaging

Packaged under Argon.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service