Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8904

Sigma-Aldrich

4-(Dimethylamino)benzaldehyde

ACS reagent

Synonym(s):

Ehrlich’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CHO
CAS Number:
Molecular Weight:
149.19
Beilstein/REAXYS Number:
606802
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

ACS reagent

impurities

Color of hydrochloric acid solution, passes test

ign. residue

≤0.1%

mp

72-75 °C (lit.)
73-75 °C (lit.)

solubility

alcohol: soluble (APHA ≤60)
hydrochloric acid: soluble

SMILES string

CN(C)c1ccc(C=O)cc1

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Forms colored condensation products (Schiff bases) with pyrroles and primary amines.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service