Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8875

Sigma-Aldrich

2-Deoxy-D-glucose 6-phosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O8P · xNa+
CAS Number:
Molecular Weight:
244.14 (free acid basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

[Na].OC(COP(O)(O)=O)C(O)C(O)CC=O

InChI

1S/C6H13O8P.Na.H/c7-2-1-4(8)6(10)5(9)3-14-15(11,12)13;;/h2,4-6,8-10H,1,3H2,(H2,11,12,13);;

InChI key

NBDKYLFTCAOTAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service