Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D7142

Sigma-Aldrich

3,2′-Dimethyl-4-aminobiphenyl hydrochloride

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N · HCl
CAS Number:
Molecular Weight:
233.74
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥95% (HPLC)

SMILES string

Cl.Cc1cc(ccc1N)-c2ccccc2C

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Shirai et al.
Mutation research, 506-507, 129-136 (2002-09-28)
2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP), the most abundant carcinogenic heterocyclic amine in cooked meat and fish, is speculated to be associated with human carcinogenesis. It has been shown to induce DNA adducts in a variety of organs in rodents and thus clarification of
K Kato et al.
Japanese journal of cancer research : Gann, 91(8), 786-791 (2000-08-31)
High intake of phytoestrogens through soybeans and their products is thought to be associated with low incidences of prostate and / or breast cancer in Asian countries. Possible chemopreventive effects of genistin or daidzin on rat prostate carcinogenesis were therefore
T Shirai et al.
Mutation research, 462(2-3), 219-226 (2000-04-18)
A number of rodent models of prostate carcinoma development have been established to study mechanisms and modifying potential. All except for transgenic mouse models need long experimental periods for generation of a high yield of cancers. Spontaneous prostate tumor models
Hiroyuki Kohno et al.
Ecotoxicology and environmental safety, 51(3), 210-215 (2002-04-25)
The modifying effects of dietary feeding of an estrogenic compound 4-n-octylphenol on 3,2'-dimethyl-4-aminobiphenyl (DMAB) induced prostatic carcinogenesis were investigated in male F344 rats. The authors also assessed the effects of test compound on proliferating cell nuclear antigen (PCNA) index in
Toshio Ichihara et al.
The Journal of toxicological sciences, 28(3), 165-171 (2003-09-17)
The current study was designed to examine the modulating effects of bisphenol A (BPA) on prostate cancer risk in male offspring exposed transplacentally and lactationally. BPA was administered to F344 female rats by gavage at 0, 0.05, 7.5, 30, 120

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service