Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D5907

Sigma-Aldrich

Dicarboxidine dihydrochloride

Synonym(s):

γ,γ′-(4,4′-Diamino-3,3′-biphenylylenedioxy)dibutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O6 · 2HCl
CAS Number:
Molecular Weight:
461.34
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

SMILES string

Cl.Nc1ccc(cc1OCCCC(O)=O)-c2ccc(N)c(OCCCC(O)=O)c2

Substrates

Chromogen for detecting N-t-BOC amino acids, peptides and barbiturates by TLC; alternative to conventional benzidine reagents as a peroxidase substrate.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of plasma hemoglobin using dicarboxidine.
B Swolin et al.
Clinica chimica acta; international journal of clinical chemistry, 121(3), 389-391 (1982-06-03)
Dicarboxidine [gamma, gamma'-(4,4'-diamino-3,3'-biphenylylenedioxy)dibutyric acid] dihydrochloride as a chromogen for the detection of barbiturates, meprobamate and some other sedatives on thin-layer chromatograms.
C M Svahn et al.
Journal of chromatography, 170(1), 294-295 (1979-02-11)
The assay of peroxidases by means of dicarboxidine on enzyme-linked immunosorbent assay level.
K G Paul et al.
Analytical biochemistry, 124(1), 102-107 (1982-07-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service