Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D5651

SAFC

(−)-N,N′-Dibenzyl-D-tartaric diamide

Synonym(s):

(−)-N,N′-Dibenzyl-D-tartramide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH(OH)CONHCH2C6H5]2
CAS Number:
Molecular Weight:
328.36
Beilstein/REAXYS Number:
4551861
MDL number:
UNSPSC Code:
12352200

mp

198-200 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(=O)NCc1ccccc1)C(=O)NCc2ccccc2

InChI

1S/C18H20N2O4/c21-15(17(23)19-11-13-7-3-1-4-8-13)16(22)18(24)20-12-14-9-5-2-6-10-14/h1-10,15-16,21-22H,11-12H2,(H,19,23)(H,20,24)/t15-,16-/m0/s1

InChI key

BBYSAHVLSFBCMN-HOTGVXAUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service