Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D5649

Sigma-Aldrich

Demissidine

Synonym(s):

Solanine D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45NO
CAS Number:
Molecular Weight:
399.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@@]4(C)[C@@]3([H])C[C@]5([H])N6C[C@@H](C)CC[C@]6([H])[C@@H](C)[C@]45[H])[C@@]1(C)CC[C@H](O)C2

Other Notes

Alkaloid found, as various glycosides, in the skin of unripe potatoes. Inhibits growth of human colon and liver cancer cells in culture, though less actively than its glycosides.

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kap-Rang Lee et al.
Journal of agricultural and food chemistry, 52(10), 2832-2839 (2004-05-13)
As part of an effort to improve plant-derived foods such as potatoes, eggplants, and tomatoes, the antiproliferative activities against human colon (HT29) and liver (HepG2) cancer cells of a series of structurally related individual compounds were examined using a microculture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service