D5288
3′-Deoxythymidine 5′-triphosphate trilithium salt
aqueous solution (10 mM, pH 7.0)
Synonym(s):
2′,3′-Dideoxythymidine 5′-triphosphate trilithium salt, dTTP Li3, ddTTP-Li3
About This Item
form
aqueous solution (10 mM, pH 7.0)
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Li+].[Li+].[Li+].CC1=CN([C@H]2CC[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O2)C(=O)NC1=O
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service