Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4023

Sigma-Aldrich

N,N-Diethyl-p-phenylenediamine sulfate salt

Synonym(s):

4-Amino-N,N-diethylaniline sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4NH2 · H2SO4
CAS Number:
Molecular Weight:
262.33
Beilstein/REAXYS Number:
4219768
EC Number:
MDL number:
UNSPSC Code:
12171500

mp

184-186 °C (lit.)

suitability

suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils

SMILES string

OS(O)(=O)=O.CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2.H2O4S/c1-3-12(4-2)10-7-5-9(11)6-8-10;1-5(2,3)4/h5-8H,3-4,11H2,1-2H3;(H2,1,2,3,4)

InChI key

AYLDJQABCMPYEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masoumeh Hasani et al.
Journal of hazardous materials, 157(1), 161-169 (2008-02-15)
A multicomponent analysis method based on principal component analysis-artificial neural network models (PC-ANN) is proposed for the determination of phenolic compounds. The method relies on the oxidative coupling of phenols (phenol, 2 chlorophenol, 3-chlorophenol and 4-chlorophenol) to N,N-diethyl-p-phenylenediamine in the
Allergic contact dermatitis in a photographer.
A Aguirre et al.
Contact dermatitis, 27(5), 340-341 (1992-11-01)
Jessica A Kershaw et al.
Analytical and bioanalytical chemistry, 379(4), 707-713 (2004-06-04)
The effect of Cu(II) on the determination of homocysteine via its electrochemically initiated reaction with N, N-diethyl-p-phenylenediamine is examined. The presence of copper inhibited the detection process for homocysteine owing to a complexation reaction occurring. This provided an indirect route
Z B Ogel et al.
Applied microbiology and biotechnology, 71(6), 853-862 (2006-01-04)
Scytalidium thermophilum produces an extracellular phenol oxidase on glucose-containing medium. Certain phenolic acids, specifically gallic acid and tannic acid, induce the expression of the enzyme. Production at 45 degrees C in batch cultures is growth-associated and is enhanced in the
Measurement of the effect of a plant-based diet on reactive oxygen metabolite production using the d-Roms test is invalid.
M I Harma et al.
The International journal of biological markers, 21(1), 60-60 (2006-05-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service