Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D155

Supelco

Dihydroergocristine methanesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H41N5O5 · CH4O3S
CAS Number:
Molecular Weight:
707.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: 10 mg/mL
methanol: 22 mg/mL

application(s)

forensics and toxicology
veterinary

SMILES string

CS(O)(=O)=O.[H][C@@]12Cc3c[nH]c4cccc(c34)[C@@]1([H])C[C@H](CN2C)C(=O)N[C@@]5(O[C@]6(O)N([C@@H](Cc7ccccc7)C(=O)N8CCC[C@@]68[H])C5=O)C(C)C

InChI

1S/C35H41N5O5.CH4O3S/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34;1-5(2,3)4/h4-7,9-12,18,20,23,25,27-29,36,44H,8,13-17,19H2,1-3H3,(H,37,41);1H3,(H,2,3,4)/t23-,25-,27-,28+,29+,34-,35+;/m1./s1

InChI key

SPXACGZWWVIDGR-SPZWACKZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ergot alkaloid. Vasoconstrictor; and partial agonist at dopaminergic and adrenergic receptors; antagonist at serotonergic receptors.

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Crystepin CH. A multi-centre trial of an antihypertensive drug (author's transl)].
O Mayer et al.
Casopis lekaru ceskych, 120(34), 1030-1037 (1981-08-27)
Hyung Gon Lee et al.
Yonsei medical journal, 51(6), 960-964 (2010-09-30)
The phosphodiesterase 5 inhibitor sildenafil has antinociceptive effects, mediated by an increase in cGMP. This study examined the role of spinal adenosine and serotonin receptors played in the antinociceptive effects of intrathecal sildenafil. Intrathecal catheters were inserted into the subarachnoid
Beatriz Bicalho et al.
Current drug metabolism, 6(6), 519-529 (2005-12-29)
Dihydroergocristine (DHEC) is a semi-synthetic drug mainly used for age-related cognitive impairment. In this study, its major metabolite 8'-hydroxy-dihydroergocristine (8'-OH-DHEC) was produced in incubates of a bovine liver preparation using dihydroergocristine mesylate (DHECM) as substrate. Purification was achieved by flash
Myung Ha Yoon et al.
Journal of Korean medical science, 18(2), 255-261 (2003-04-15)
Spinal gabapentin has been known to show the antinociceptive effect. Although several assumptions have been suggested, mechanisms of action of gabapentin have not been clearly established. The present study was undertaken to examine the action mechanisms of gabapentin at the
Michal Dousa et al.
Journal of AOAC International, 93(1), 97-101 (2010-03-26)
A rapid procedure based on a direct extraction and HPLC determination of dihydroergocristine in a pharmaceutical preparation with fluorescence detection has been developed and validated. The optimized chromatographic conditions included a Purospher RP18e column, 5 microm particle size, 250 x

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service