D1400
Deoxycorticosterone 21-hemisuccinate
Synonym(s):
21-Hydroxy-4-pregnene-3,20-dione 21-hemisuccinate, 21-Hydroxyprogesterone 21-hemisuccinate, 4-Pregnen-21-ol-3,20-dione 21-hemisuccinate, Cortexone 21-hemisuccinate
About This Item
storage temp.
−20°C
SMILES string
[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])C(=O)COC(=O)CCC(O)=O
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service