Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1265

Sigma-Aldrich

Diethylene glycol monoethyl ether

Synonym(s):

2-(2-Ethoxyethoxy)ethanol, CARBITOL, Diethylene glycol ethyl ether, Ethyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein/REAXYS Number:
1736441
EC Number:
MDL number:
UNSPSC Code:
12352201

vapor density

4.63 (vs air)

vapor pressure

0.12 mmHg ( 20 °C)

expl. lim.

1.2 %, 135 °F
23.5 %, 182 °F

refractive index

n20/D 1.427 (lit.)

bp

202 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCO

InChI

1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3

InChI key

XXJWXESWEXIICW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service