Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1022

Sigma-Aldrich

3,5-Diamino-1,2,4-triazole

powder

Synonym(s):

Guanazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5N5
CAS Number:
Molecular Weight:
99.09
Beilstein/REAXYS Number:
112467
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

202-205 °C (lit.)

solubility

H2O: soluble, clear to hazy

application(s)

forensics and toxicology
veterinary

SMILES string

Nc1n[nH]c(N)n1

InChI

1S/C2H5N5/c3-1-5-2(4)7-6-1/h(H5,3,4,5,6,7)

InChI key

PKWIYNIDEDLDCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service