Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0909

Sigma-Aldrich

DEAE-Cellulose

fibers

Synonym(s):

Cellulose DEAE, Diethylaminoethyl-cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
PubChem Substance ID:

form

fibers

capacity

~1.0 meq/g small ion capacity

SMILES string

OC[C@@H]1O[C@@H](O[C@H]2[C@@H](O)[C@H](O)[C@@H](O)O[C@H]2CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John S Scott-Craig et al.
The Journal of biological chemistry, 286(50), 42848-42854 (2011-10-29)
α-Linked xylose is a major component of xyloglucans in the cell walls of higher plants. An α-xylosidase (AxlA) was purified from a commercial enzyme preparation from Aspergillus niger, and the encoding gene was identified. The protein is a member of
Tiago Vicente et al.
Biotechnology and bioengineering, 108(6), 1347-1359 (2011-02-05)
The effect of ligand density on anion-exchange membrane chromatography (AEXmc) for the purification of recombinant baculoviruses (rBVs), potential viral vectors in clinical applications, is studied by surface plasmon resonance on customized AEX surfaces and gradient elution experiments on Sartobind D
S A Glushkov et al.
Analytical biochemistry, 393(1), 135-137 (2009-06-16)
Polymerase chain reaction (PCR) is widely used to detect specific DNA sequences for purposes of microbial identification, clinical diagnosis, and basic research. The most pernicious problem plaguing this technique is contamination of PCR reagents with previously amplified material. We propose
Jack H Wong et al.
Journal of the science of food and agriculture, 90(1), 70-77 (2010-04-01)
A haemagglutinin was purified from Japanese Hokkaido red beans (Phaseolus vulgaris cv. Hokkaido red bean) with a procedure that included three chromatographic media. Haemagglutinating activity was adsorbed on DEAE cellulose, Affi-gel blue gel and Mono S. The pure haemagglutinin was
D Z Ma et al.
Peptides, 30(12), 2089-2094 (2009-09-02)
An antifungal defensin-like peptide with a molecular mass of 7.1kDa was isolated from dried Nepalese large red beans (Phaseolus angularis). The purification protocol employed included ion exchange chromatography on DEAE-cellulose, affinity chromatography on Affi-gel blue gel, ion exchange chromatography on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service