Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0625

Sigma-Aldrich

N,N′-Didansyl-L-cystine

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H34N4O8S4
CAS Number:
Molecular Weight:
706.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95%

storage temp.

−20°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NC(CSSCC(NS(=O)(=O)c3cccc4c(cccc34)N(C)C)C(O)=O)C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service