Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0554

Sigma-Aldrich

3,6-Dihydroxyphthalonitrile

Synonym(s):

2,3-Dicyano-1,4-hydroquinone, 2,3-Dicyanohydroquinone, DCH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4N2O2
CAS Number:
Molecular Weight:
160.13
Beilstein/REAXYS Number:
2101249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

SMILES string

Oc1ccc(O)c(C#N)c1C#N

InChI

1S/C8H4N2O2/c9-3-5-6(4-10)8(12)2-1-7(5)11/h1-2,11-12H

InChI key

MPAIWVOBMLSHQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service