Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9889

Sigma-Aldrich

Chromosorb® W/AW-DMCS

High performance, 100-120 mesh

Synonym(s):

Diatomaceous earth, flux-calcined w/DMCS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

particle size

100-120 mesh

density

0.18 g/cm3 (loose weight)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

Inchi Key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acid washed and treated with dimethyldichlorosilane

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service