Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9653

Sigma-Aldrich

(−)-trans-Caryophyllene

Synonym(s):

β-Caryophyllene, trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein/REAXYS Number:
2044564
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

bp

262-264 °C (lit.)

density

0.902 g/mL at 20 °C (lit.)

SMILES string

[H][C@@]12CC\C(C)=C\CCC(=C)[C@H]1CC2(C)C

InChI

1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6+/t13-,14-/m1/s1

InChI key

NPNUFJAVOOONJE-GFUGXAQUSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

221.9 °F - closed cup

flash_point_c

105.5 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Zhai et al.
Journal of medical entomology, 54(3), 658-669 (2017-04-12)
The roots of Eleutherococcus senticosus (Rupr. & Maxim.) Maxim., a well-known medicinal plant from Eastern Asia, are used worldwide for their known beneficial medicinal properties. Recently, the leaves have been used as an alternative to the roots. The present study
Louis M T Bradbury et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(27), E1888-E1897 (2012-06-19)
In photosynthetic organisms, carotenoids serve essential roles in photosynthesis and photoprotection. A previous report designated CruP as a secondary lycopene cyclase involved in carotenoid biosynthesis [Maresca J, et al. (2007) Proc Natl Acad Sci USA 104:11784-11789]. However, we found that
Pablinny Moreira Galdino et al.
Progress in neuro-psychopharmacology & biological psychiatry, 38(2), 276-284 (2012-05-01)
Spiranthera odoratissima A. St. Hil. (manacá) is used in folk medicine to treat renal and hepatic diseases, stomachache, headaches and rheumatism. A central nervous system (CNS) depressant effect of the hexane fraction from the ethanolic extract of this plant has
Rajesh K Joshi
Natural product communications, 8(3), 401-402 (2013-05-18)
The essential oil composition from the aerial parts of Baccharoides lilacina (Dalzell & A. Gibson) M. R. Almeida was analyzed by gas chromatography (GC) and gas chromatography/mass spectrometry (GC/MS). A total of 41 compounds have been identified, representing 97.4% of
Rajesh K Joshi
Natural product communications, 8(2), 225-226 (2013-03-22)
The hydro-distilled essential oil of dried flowers of Anaphalis contorta Hook f. has been examined by means of gas chromatography-mass spectrometry (GC-MS). The oil constituents were identified according to their mass spectra and their relative retention indices determined on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service