Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9014

Supelco

Chromosorb® G

AW-DMCS, 80-100 mesh particle size, bottle of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

packaging

bottle of 100 g

particle size

80-100 mesh

density

0.47 g/cm3 (loose weight)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acid washed and treated with dimethyldichlorosilane

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service