Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8270

Sigma-Aldrich

Cephapirin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H16N3O6S2Na
CAS Number:
Molecular Weight:
445.45
EC Number:
MDL number:
UNSPSC Code:
12352200

mode of action

cell wall synthesis | interferes

Storage temp.

2-8°C

SMILES string

[Na].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CSc3ccncc3)C2=O)C(O)=O

General description

Chemical structure: ß-lactam

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyson R Ribeiro et al.
Chemosphere, 169, 524-533 (2016-11-30)
Cefapirin (CEPA) and ceftiofur (CEF) are two examples of widely used veterinarian cephalosporins presenting multiple ionization centers. However, the acid dissociation constants (pK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service