Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8162

Supelco

Cocaine-d3

Synonym(s):

3-Benzoyloxy-8-(methyl-d3)-8-azabicyclo[3.2.1]octane-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17D3H18NO4
CAS Number:
Molecular Weight:
306.37
MDL number:
UNSPSC Code:
41116107

isotopic purity

98 atom % D

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

[2H]C([2H])([2H])N1C2CCC1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(=O)OC

Application

Cocaine-d3 may be used as an internal standard for the determination of cocaine in urine samples using solid phase microextraction (SPME) combined with direct analysis in real time (DART) coupled with tandem mass spectrometry.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of cocaine and methadone in urine samples by thin-film solid-phase microextraction and direct analysis in real time (DART) coupled with tandem mass spectrometry.
Rodriguez-Lafuente A, et al.
Analytical and Bioanalytical Chemistry, 405(30), 9723-9727 (2013)
S P Jindal et al.
Journal of pharmaceutical sciences, 67(6), 811-814 (1978-06-01)
A quantitative GLC-mass spectrometric assay was developed for the determination of cocaine and its principal metabolite, benzoylecgonine, in human urine. The assay utilizes selective ion focusing to monitor in a GLC effluent the molecular ions of cocaine and benzoylecgonine generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service