Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6862

Sigma-Aldrich

CB 34

>98% (HPLC)

Synonym(s):

N,N-Dipropyl-2-(4-chlorophenyl)-6,8-dichloro-imidazo[1,2-a]pyridine-3-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N3Cl3O
CAS Number:
Molecular Weight:
438.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 26 mg/mL
H2O: insoluble

SMILES string

CCCN(CCC)C(=O)Cc1c(nc2c(Cl)cc(Cl)cn12)-c3ccc(Cl)cc3

InChI

1S/C21H22Cl3N3O/c1-3-9-26(10-4-2)19(28)12-18-20(14-5-7-15(22)8-6-14)25-21-17(24)11-16(23)13-27(18)21/h5-8,11,13H,3-4,9-10,12H2,1-2H3

InChI key

QYVWBXJKZUCSNI-UHFFFAOYSA-N

Gene Information

human ... BZRAP1(9256)
rat ... Tspo(24230)

Biochem/physiol Actions

Selective high affinity ligand at peripheral benzo­diazepine receptors.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service