C6051
Chromium(VI) oxide
ACS reagent, ≥98.0%
Synonym(s):
Chromic anhydride
About This Item
Recommended Products
grade
ACS reagent
assay
≥98.0%
impurities
≤0.01% Insoluble matter
mp
196 °C (dec.) (lit.)
anion traces
chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤0.05%
sulfate (SO42-): ≤0.005%
cation traces
Fe, Al, Ba: ≤0.03%
Na: ≤0.2%
SMILES string
O=[Cr](=O)=O
InChI
1S/Cr.3O
InChI key
WGLPBDUCMAPZCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3
target_organs
Respiratory system
Storage Class
5.1A - Strongly oxidizing hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service