All Photos(1)
About This Item
Linear Formula:
C10H18(=O)
CAS Number:
Molecular Weight:
154.25
Beilstein/REAXYS Number:
1617127
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
refractive index
n20/D 1.482 (lit.)
bp
106-107 °C/12 mmHg (lit.)
mp
21-24 °C (lit.)
density
0.958 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
O=C1CCCCCCCCC1
InChI
1S/C10H18O/c11-10-8-6-4-2-1-3-5-7-9-10/h1-9H2
InChI key
SXOZDDAFVJANJP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service