Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3490

Sigma-Aldrich

Cyclofenil

≥98% (HPLC), white, solid

Synonym(s):

4,4′-Cyclohexylidenemethylenediphenol, 4-(Cyclohexylidenemethylene)-bis-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O4
CAS Number:
Molecular Weight:
364.43
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(cc1)\C(=C2\CCCCC2)c3ccc(OC(C)=O)cc3

InChI

1S/C23H24O4/c1-16(24)26-21-12-8-19(9-13-21)23(18-6-4-3-5-7-18)20-10-14-22(15-11-20)27-17(2)25/h8-15H,3-7H2,1-2H3

InChI key

GVOUFPWUYJWQSK-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Cyclofenil is a selective estrogen receptor β antagonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Rossi et al.
The Italian journal of gastroenterology, 24(2), 77-78 (1992-02-01)
The case of a 47-year old woman suffering from acute hepatitis caused by cyclofenil, a drug proposed for the treatment of anovulation and scleroderma, is presented. Hepatitis developed seven weeks after the beginning of administration of the drug and its
Peter Gärtner et al.
Journal of mass spectrometry : JMS, 43(7), 958-964 (2008-06-26)
The detection of metabolites of the anti-estrogenic substance cyclofenil, listed on the World Anti-Doping Agency (WADA) Prohibited List since 2004 is described. Target substances are hydroxylated metabolites, bearing an aliphatic hydroxyl group either in the 2-, 3- or 4-position of
Infertility and subfertility.
Kirsten Duckitt
Clinical evidence, (11)(11), 2427-2458 (2005-01-18)
Karen J Kieser et al.
Journal of medicinal chemistry, 53(8), 3320-3329 (2010-03-26)
Selective estrogen receptor (ER) down-regulators (SERDs) reduce ERalpha protein levels as well as block ER activity and therefore are promising therapeutic agents for the treatment of hormone refractory breast cancer. Starting with the triarylethylene acrylic acid SERD 4, we have
D Drecktrah et al.
Journal of cell science, 111 ( Pt 7), 951-965 (1998-05-20)
Previous studies have shown that the Golgi stack and the trans-Golgi network (TGN) may play a role in capturing escaped resident endoplasmic reticulum (ER) proteins, and directing their retrograde transport back to that organelle. Whether this retrograde movement represents a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service