C2661
Calcium chloride
suitable for plant cell culture, anhydrous
About This Item
Recommended Products
grade
anhydrous
vapor pressure
0.01 mmHg ( 20 °C)
technique(s)
cell culture | plant: suitable
mp
772 °C (lit.)
SMILES string
[Cl-].[Cl-].[Ca++]
InChI
1S/Ca.2ClH/h;2*1H/q+2;;/p-2
InChI key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service