Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2604

Sigma-Aldrich

Compound 52

≥98.0%, powder

Synonym(s):

2-(2-Hydroxyethylamino)-6-(3-chloroanilino)-9-isopropylpurine, NG-52

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN6O
CAS Number:
Molecular Weight:
346.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0%

form

powder

color

white to off-white

solubility

methylene chloride: 50 mg/mL, clear, colorless

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)n1cnc2c(Nc3cccc(Cl)c3)nc(NCCO)nc12

Biochem/physiol Actions

2,6,9-substituted purine inhibitor of cyclin-dependent kinases.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Muta. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Liang Wang et al.
Acta pharmacologica Sinica, 42(4), 633-640 (2020-08-02)
Inhibition of glycolysis process has been an attractive approach for cancer treatment due to the evidence that tumor cells are more dependent on glycolysis rather than oxidative phosphorylation pathway. Preliminary evidence shows that inhibition of phosphoglycerate kinase 1 (PGK1) kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service