Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2536

Sigma-Aldrich

Calcium chloride dihydrate

suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98%

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

mp

176 °C (dec.) (lit.)

solubility

H2O: 294 mg/mL, clear, colorless

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Geetha et al.
Ecotoxicology and environmental safety, 122, 17-23 (2015-07-15)
Population explosion, depletion of water resources and prolonged droughts and floods due to climatic change lead to scarcity of pure and hygienic drinking water in most of the developing countries. Recently nanomaterials attained considerable attention as biosorbent for water purification
Avinash A Kadam et al.
Bioresource technology, 132, 276-284 (2013-02-16)
Pretreatments to sugarcane bagasse (SCB) such as CaCl2, alkali, ammonia, steam and milling showed 91%, 46%, 47%, 42% and 56% adsorption of Solvent Red 5B (SR5B); 92%, 57%, 58%, 56% and 68% adsorption of simulated dyes mixture (SDM), and 86%
Robert B Hamanaka et al.
Science signaling, 6(261), ra8-ra8 (2013-02-07)
Proper regulation of keratinocyte differentiation within the epidermis and follicular epithelium is essential for maintenance of epidermal barrier function and hair growth. The signaling intermediates that regulate the morphological and genetic changes associated with epidermal and follicular differentiation remain poorly
Meral Tunc-Ozdemir et al.
Plant physiology, 161(2), 1010-1020 (2013-02-02)
Cyclic nucleotide-gated channels (CNGCs) have been implicated in diverse aspects of plant growth and development, including responses to biotic and abiotic stress, as well as pollen tube growth and fertility. Here, genetic evidence identifies CNGC16 in Arabidopsis (Arabidopsis thaliana) as
Marianne T Vestermark et al.
Acta orthopaedica, 82(5), 614-621 (2011-09-08)
Allografts are often used during revision hip replacement surgery for stabilization of the implant. Resorption of the allograft may exceed new bone formation, and instability of the prosthesis can develop. We investigated whether strontium could regulate the imbalance of fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service