Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1283

Sigma-Aldrich

Z-D-Leu-OH

Synonym(s):

N-Cbz-D-leucine, Z-D-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Beilstein/REAXYS Number:
4354627
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17)/t12-/m1/s1

InChI key

USPFMEKVPDBMCG-GFCCVEGCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service