C0306
7-Chlorokynurenic acid
powder
Synonym(s):
7-Chloro-4-hydroxyquinoline-2-carboxylic acid, 7-Cl-KYNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H6ClNO3
CAS Number:
Molecular Weight:
223.61
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
form
powder
SMILES string
OC(=O)c1cc(O)c2ccc(Cl)cc2n1
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Potent NMDA glutamate receptor antagonist; antagonizes the strychnine-insensitive glycine site of the NMDA receptor.
Potent NMDA glutamate receptor antagonist; antagonizes the strychnine-insensitive glycine site of the NMDA receptor; prevents neurodegeneration produced by quinolinic acid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service