All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H16N5O7P
CAS Number:
Molecular Weight:
433.31
UNSPSC Code:
12352204
storage temp.
−20°C
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kevin W-H Lo et al.
Journal of tissue engineering and regenerative medicine, 6(1), 40-48 (2011-02-12)
Osteoblastic differentiation is an important landmark for bone formation, bone repair and regeneration; however, it is a very complex process controlled by different signalling mechanisms. Several groups have reported that the cyclic adenosine monophosphate (cAMP) signalling system is responsible for
R Bøe et al.
British journal of cancer, 72(5), 1151-1159 (1995-11-01)
8-Cl-cAMP and 8-NH2-cAMP induced MCF-7 cell death. The type(s) of cell death were studied in more detail and compared with the cell death type (apoptosis) induced by okadaic acid, an inhibitor of serine/threonine phosphatases. By morphological criteria dying cells showed
N Saati et al.
In vitro cellular & developmental biology. Animal, 33(4), 310-314 (1997-04-01)
Agents that increase intracellular cAMP (cAMP elevating agents) and 1, 25(OH)2D3 inhibit the proliferation of many cell types. We investigated the combined effect of 1,25(OH)2D3 and cAMP elevating agents on exponentially growing mouse 3T3 fibroblasts. The following cAMP elevating agents
W Eskild et al.
Biochemical and biophysical research communications, 152(3), 1504-1510 (1988-05-16)
The levels of mRNA for cellular retinol binding protein (CRBP) were studied in primary rat Sertoli cell cultures treated with cAMP analogues and retinol. In the presence of cyclic AMP analogues a dose- and time-dependent reduction (70-90%) of the levels
R A Steinberg et al.
The Journal of biological chemistry, 271(44), 27630-27636 (1996-11-01)
The guanidinium groups of conserved arginines in the two intrachain cAMP-binding sites of regulatory (R) subunit of cAMP-dependent protein kinase have been implicated in the allosteric interactions by which cAMP binding leads to kinase activation. We have investigated the functional
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service