Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6383

Sigma-Aldrich

9-Bbn dimer

Synonym(s):

9-Borabicyclo[3.3.1]nonane dimer, 9-BBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30B2
CAS Number:
Molecular Weight:
244.03
Beilstein/REAXYS Number:
605509
MDL number:
UNSPSC Code:
12000000

mp

150-152 °C (lit.)

storage temp.

2-8°C

SMILES string

C1C[C@H]2CCC[C@@H](C1)B2B3[C@H]4CCC[C@@H]3CCC4

InChI

1S/C16H28B2/c1-5-13-7-2-8-14(6-1)17(13)18-15-9-3-10-16(18)12-4-11-15/h13-16H,1-12H2/t13-,14+,15-,16+

InChI key

IYDIZBOKVLHCQZ-GEEKYZPCSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service