B3791
B-Pentasaccharide
from human urine, ≥85%
Synonym(s):
α-Fuc-(1→2)-α-Gal-(1→3)-β-Gal-(1→4)(α-Fuc-[1→3])-Glc
Select a Size
Select a Size
About This Item
biological source
human urine
assay
≥85%
form
powder
storage temp.
2-8°C
SMILES string
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)C(O)O[C@H](CO)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H]3O[C@@H]5O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]5O)[C@@H](O)[C@H](O)[C@@H]1O
InChI
1S/C30H52O24/c1-6-11(34)15(38)18(41)27(46-6)53-24-21(44)26(45)48-10(5-33)22(24)51-30-25(54-28-19(42)16(39)12(35)7(2)47-28)23(14(37)9(4-32)50-30)52-29-20(43)17(40)13(36)8(3-31)49-29/h6-45H,3-5H2,1-2H3/t6-,7-,8+,9+,10+,11+,12+,13-,14-,15+,16+,17-,18-,19-,20+,21+,22+,23-,24+,25+,26?,27-,28-,29+,30-/m0/s1
InChI key
CFAMAWVZEVLFJH-NCWFDBPASA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service