Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3504

Sigma-Aldrich

Phenylglyoxylic acid

powder

Synonym(s):

α-Oxophenylacetic acid, Benzoylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCOOH
CAS Number:
Molecular Weight:
150.13
Beilstein/REAXYS Number:
606718
EC Number:
MDL number:
UNSPSC Code:
12352100

form

powder

color

pale yellow to pink

mp

62-65 °C (lit.)

SMILES string

OC(=O)C(=O)c1ccccc1

InChI

1S/C8H6O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5H,(H,10,11)

InChI key

FAQJJMHZNSSFSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service